Learn More
Sigma Aldrich Fine Chemicals Biosciences C1 Esterase inhibitor from1MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SRP63171MG
Encompass
C1 esterase inhibitor is a single chain glycoprotein which inhibits C1 C1r C1s plasma kallikrein factors XIa XIIa and plasmin of the blood clotting system. It is present in the plasma at 16-33 mg/100mL. It is part of the serpin family.
Catalog No. 50-290-6590
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.