Learn More
Vee Gee Vee Gee Abbe Refractometer
Supplier: Vee Gee C10
The C10 Abbe Refractometer is a high precision benchtop instrument measuring the Refractive Index (nD) of liquids, glass, plastic and film. With an oversized, easy to read scale and Prism Temperature Control feature, the C10 delivers Brix and Refractive Index Scales and dispersion measurements with high precision. Dispersion (nF-nC): Dial Readings w/Conversion Charts Ambient Temperature: 5 to 35 C Measuring Temperature: 0 to 70 C Optical Wavelength: 589 nm Prism: Optical Glass Prism Housing: SUS316 Stainless Steel Circulator Nozzles: 2 for Primary Prism, 2 for Secondary Prism; accepts 3/8"""" ID tubing Dimensions: 165 (L) x 88.9 (W) x 238.8 (H) mm (6.5 x 3.5 x 9.4), Weight: 4.5 kg (9.9 lbs.)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.