Learn More
AdipoGen C10-DL-HSL
SDP
Supplier:  AdipoGen CDXD0338M050
Encompass_Preferred
Chemical. CAS 106983-36-2. Formula C14H25NO3. MW 255.4. Synthetic. N-Decanoyl-DL-homoserine lactone N-C10-HSL is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-256-1103
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.