Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cortisol Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco C1061ML

Catalog No. 01-001-072


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Also known as hydrocortisone, cortisol is a glucocorticoid produced by the adrenal gland. The hormone is measured in whole blood by LC-MS/MS for newborn screening of congenital adrenal hyperplasia (CAH) and in serum or plasma by LC-MS/MS for determination of several diseases including Cushing's syndrome, Addison's disease, and Sheehan's syndrome.
Specifications

Specifications

67-56-1, 50-23-7
UN 1230 - class 3 - PG 2 - Methanol, solution
Hydrocortisone solution, 17-Hydroxycorticosterone, Cortisol
1 mL
MFCD00011654
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

CERILLIANT is a registered trademark of Cerilliant Corporation