Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf71 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546316 Supplier Invitrogen™ Supplier No. PA546316
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AERSSYENKE VEGELEMGPA GSSWCPDSRE HRPRKHLSLR LCNRDPEPGG Sequence homology: Human: 100%.

Plays an important role in cardiomyocyte hypertrophy via activation of the calcineurin/NFAT signaling pathway. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf71
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C10orf71
Gene Accession No. Q711Q0
Gene Alias C10orf71; Cardiac-enriched FHL2-interacting protein; CEFIP; chromosome 10 open reading frame 71; uncharacterized protein C10orf71
Gene Symbols C10orf71
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human C10orf71 (aa 609-658).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118461
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.