Learn More
C11orf79 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
19906-1-AP is a rabbit polyclonal antibody raised against the full-length of human SDHAF2.
Specifications
Specifications
| Antigen | C11orf79 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.29 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | SDHAF2 |
| Gene Accession No. | Q5RJQ7, Q8C6I2, Q9NX18 |
| Gene Alias | C11orf79, hSDH5, PGL2, SDH assembly factor 2, SDH5, SDHAF2, SDHAF2, PGL2, SDH5 |
| Gene Symbols | Sdhaf2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.