Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113555
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113555 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113555 Supplier Invitrogen™ Supplier No. PA5113555
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C11orf9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

MYRF is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MYRF
Gene Accession No. Q3UR85, Q9Y2G1
Gene Alias 6030439E18; C11orf9; C18H11orf9; Gm1804; Gm98; KIAA0954; Mrf; myelin gene regulatory factor; myelin gene regulatory factor; myelin regulatory factor; myelin gene regulatory factor-like; myelin regulatory factor; Myelin regulatory factor, C-terminal; Myelin regulatory factor, N-terminal; MYRF; Ndt80; pqn-47; RGD1306622; zgc:172183
Gene Symbols MYRF
Host Species Rabbit
Immunogen Recombinant Human Myelin regulatory factor protein (843-1004AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225908, 745
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.