Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf94 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570982 Supplier Invitrogen™ Supplier No. PA570982
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 79%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 93%; Rat: 86%.

C11orf94 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf94
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C11orf94
Gene Accession No. C9JXX5
Gene Alias C11orf94; chromosome 11 open reading frame 94; uncharacterized protein C11orf94
Gene Symbols C11orf94
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C11orf94.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143678
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.