Learn More
AdipoGen C12-DL-HSL
SDP
Supplier:  AdipoGen CDXD0339M050
Encompass_Preferred
Chemical. CAS 18627-38-8. Formula C16H29NO3. MW 283.4. Synthetic. N-Dodecanoyl-DL-homoserine lactone C12-HSL is a small diffusible signalling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism in bacteria. In addition to regulating bacterial functions, C12-HSL activates NF-kappaB in RAW 264.7 macrophages, increasing the expression of TNF-alpha, interleukin-1beta IL-1beta and IL-8, while other lactones do not. In addition, C12-HSL alters cell cycling and metabolism of human keratinocyte HaCaT cells. It also was shown to induce cell death by apoptosis.
Catalog No. 50-256-1101
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.