Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576411 Supplier Invitrogen™ Supplier No. PA576411
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes the auxiliary subunit of the heteromeric N-terminal acetyltransferase B complex. This complex acetylates methionine residues that are followed by acidic or asparagine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf30
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Naa25
Gene Accession No. Q14CX7, Q6QI44, Q8BWZ3
Gene Alias 4833422K13Rik; A630046C11; AI450868; C12orf30; C330023M02Rik; Liver regeneration-related protein LRRGT00164; LRRGT00164; MDM20; mitochondrial distribution and morphology 20; mitochondrial distribution and morphology protein 20; N(alpha)-acetyltransferase 25, NatB auxiliary subunit; Naa25; Nalpha acteyltransferase 25; N-alpha-acetyltransferase 25, NatB auxiliary subunit; NAP1; NatB complex subunit MDM20; N-terminal acetyltransferase B complex subunit MDM20; N-terminal acetyltransferase B complex subunit NAA25; p120; RGD1305685
Gene Symbols Naa25
Host Species Rabbit
Immunogen Recombinant fusion protein of human NAA25.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231713, 360811, 80018
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.