Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Cortisone Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco C1301ML

Catalog No. 01-001-091


Only null left
Add to Cart

Description

Description

Cortisone is a corticosteroid produced in the adrenal glands. Cortisone is administered for short term pain relief and to reduce swelling from inflammation. This Certified Spiking Solution™ is applicable in LC-MS/MS applications for endocrinology, clinical chemistry and neonatal screening.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1, 53-06-5
UN 1230 - class 3 - PG 2 - Methanol, solution
360.44
1 mL
C21H28O5
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation