Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C13orf33 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735227 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5227 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5227 Supplier Proteintech Group Inc Supplier No. 261371AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEDAG is a protein coding gene involved in processes that promote adipocyte differentiation, lipid accumulation, and glucose uptake in mature adipocytes.

Specifications

Antigen C13orf33
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MEDAG
Gene Accession No. Q5VYS4
Gene Alias AWMS3, C13orf33, hAWMS3
Gene Symbols MEDAG
Host Species Rabbit
Immunogen C13orf33 Fusion Protein Ag23516
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84935
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.