Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf166 Mouse anti-Human, Clone: UMAB181, liquid, UltraMAB™
SDP

Catalog No. 501685523
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5523 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5523 Supplier OriGene Supplier No. UM870073
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The function of the C14orf266 gene has not yet been determined.

Specifications

Antigen C14orf166
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UMAB181
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RTRAF
Gene Accession No. Q9Y224
Gene Alias C14orf166, CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE, hCLE1
Gene Symbols RTRAF
Host Species Mouse
Immunogen Full length human recombit protein of human C14orf166 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51637
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.