Learn More
Enzo Life Sciences C16 Ceramide (5 mg). CAS: 24696-26-2
SDP
Supplier:  Enzo Life Sciences BMLSL1150005
Encompass_Preferred
An abundant molecular species of endogenous ceramide. It induces phosphorylation of epidermal growth factor receptor on Thr669 in A-431 cells by stimulation of ceramide-activated protein kinase and activates NF-κB at concentrations as low as 50 nM. Activates PKCζ. Alternative name: N-Palmitoyl-D-erythro-sphingosine, N-Hexadecanoyl-D-erythro-sphingosine, N-Palmitoyl ceramide. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (5mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-2111
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.