Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C16orf57 Monoclonal Antibody (OTI1B1), TrueMAB™, OriGene
SDP

Catalog No. 501683983
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3983 Supplier OriGene Supplier No. TA811328
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C16orf57 is a phosphodiesterase responsible for the U6 snRNA 3′ end processing. This protein acts as an exoribonuclease (RNase) responsible for trimming the poly(U) tract of the last nucleotides in the pre-U6 snRNA molecule, leading to the formation of mature U6 snRNA 3′ end-terminated with a 2′,3′-cyclic phosphate.

Specifications

Antigen C16orf57
Applications Western Blot
Classification Monoclonal
Clone OTI1B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene USB1
Gene Accession No. Q9BQ65
Gene Alias C16orf57, HVSL1, Mpn1, PN, hUsb1
Gene Symbols USB1
Host Species Mouse
Immunogen Full length human recombit protein of human C16orf57 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79650
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.