Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C16orf61 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735200 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5200 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5200 Supplier Proteintech Group Inc Supplier No. 260861AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Product No. 26086-1-AP is a rabbit polyclonal antibody raised against the full-length of human C16orf61. The MW of this protein is 9 kDa, and this antibody specifically recognizes the 9 kDa protein.

The function of C16orf61 remains largely unknown. It may be involved in cytochrome c oxidase biogenesis.

Specifications

Antigen C16orf61
Applications Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CMC2
Gene Accession No. Q9NRP2
Gene Alias 2310061C15Rik, C16orf61, CMC2, DC13
Gene Symbols CMC2
Host Species Rabbit
Immunogen C16orf61 Fusion Protein Ag23319
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56942
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.