Learn More
Epigentek Inc C17orf62 Polyclonal Antibody, Biotin Conjulated (50 ul)
SDP
Supplier:  Epigentek Inc A67153050
Encompass_Preferred
Biotin Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Cytochrome b-245 chaperone 1 protein (43-187AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Uncharacterized protein C17orf62, C17orf62.
Catalog No. 50-124-8323
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.