Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf89 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563474 Supplier Invitrogen™ Supplier No. PA563474
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSKDFCAREF EALRSCFAAA AKKTL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

NDUFAF8 is a protein coding gene involved in the assembly of mitochondrial NADH:ubiquinone oxidoreductase complex (complex I, MT-ND1). Required to stabilize NDUFAF5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf89
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFAF8
Gene Accession No. A1L188
Gene Alias 1810043H04Rik; C17orf89; C9H17orf89; chromosome 17 open reading frame 89; hypothetical protein LOC690871; LOC690871; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 8; NADH:ubiquinone oxidoreductase complex assembly factor 8; Ndufaf8; RIKEN cDNA 1810043H04 gene; uncharacterized protein C17orf89; uncharacterized protein C17orf89 homolog
Gene Symbols NDUFAF8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C17orf89. Recombinant protein control fragment (Product #RP-100584).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.