Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf96 Recombinant Rabbit Monoclonal Antibody (5H7L8)

Catalog No. 703052
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-305-2 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-305-2 Supplier Invitrogen™ Supplier No. 703052
  • $580.00
    $435.65 / Each of 1

    Save $144.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Pig, Cat, Dog. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Elongin BC and Polycomb repressive complex 2-associated protein (EPOP) also know as C17ORF96 is a scaffold protein that serves as a bridging partner between the PRC2/EED-EZH2 complex and the elongin BC complex. It is required to fine tune the transcriptional status of polycomb group (PcG) target genes in embryonic stem cells (ESCs). C17ORF96 also plays a key role in genomic regions that display both active and repressive chromatin properties in pluripotent stem cells by sustaining low level expression of PcG target genes. It acts by recruiting the elongin BC complex, thereby restricting excessive activity of the PRC2/EED-EZH2 complex. C17ORF96 also has interacts with USP7 and promotes deubiquitination of H2B at promoter sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf96
Applications ChIP Assay, Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone 5H7L8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene EPOP
Gene Accession No. A6NHQ4, Q7TNS8
Gene Alias AA409164; AI413509; AW539173; C17orf96; C9H17orf96; chromosome 17 open reading frame 96; CUNH17orf96; E13; E130012A19Rik; elongin BC and polycomb repressive complex 2 associated protein; elongin BC and Polycomb repressive complex 2-associated protein; Embryonic stem cell-specific PRC2 subunit p48; EPOP; ES cell-specific PRC2 subunit p48; esPRC2p48; hypothetical protein LOC691153; LOC691153; proline rich 28; Proline-rich protein 28; PRR28; RIKEN cDNA E130012A19 gene; uncharacterized protein C17orf96; uncharacterized protein C17orf96 homolog; Uncharacterized protein ENSP00000317905
Gene Symbols EPOP
Host Species Rabbit
Immunogen Peptides corresponding to human C17orf96 [aa130-aa148, aa360-aa379].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100170841, 103551
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.