Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf36 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735004 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5004 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5004 Supplier Proteintech Group Inc Supplier No. 256501AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IZUMO4 is a protein coding gene.

Specifications

Antigen C19orf36
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene IZUMO4
Gene Accession No. D3Z690, Q1ZYL8
Gene Alias C19orf36, IMAGE:4215339, IZUMO4, Sperm 22 kDa protein c113
Gene Symbols Izumo4
Host Species Rabbit
Immunogen C19orf36 Fusion Protein Ag20434
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113177, 71564
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.