Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf53 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563943
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563943 Supplier Invitrogen™ Supplier No. PA563943
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKNLEVGIRK KIEHDVVMKA SSSLPKKLAL LKAPAKKKGA AAATSSKTPS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 80%.

May have a potential role in hypercalcemia of malignancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf53
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C19orf53
Gene Accession No. Q9UNZ5
Gene Alias C19orf53; chromosome 19 open reading frame 53; HSPC023; Leydig cell tumor 10 kDa protein homolog; LYDG10
Gene Symbols C19orf53
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf53. Recombinant protein control fragment (Product #RP-101982).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28974
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.