Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C19orf63 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114333 Supplier Invitrogen™ Supplier No. PA5114333
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C19orf63 Polyclonal Antibody for Western Blot, IHC (P), Flow

Promotes angiogenesis and tissue repair in the heart after myocardial infarction. Stimulates cardiac endothelial cell migration and outgrowth via the activation of p38 MAPK, PAK and MAPK2 signaling pathways. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C19orf63
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene EMC10
Gene Accession No. Q3TAS6, Q5UCC4
Gene Alias 2310044H10Rik; 5430410O10Rik; C19orf63; EMC10; ER membrane protein complex subunit 10; hematopoietic signal peptide-containing membrane domain-containing 1; hematopoietic signal peptide-containing membrane domain-containing protein 1; hematopoietic signal peptide-containing secreted 1; HSM1; HSS1; Inm02; Mirta22; UNQ764/PRO1556; UPF0510 protein INM02
Gene Symbols EMC10
Host Species Rabbit
Immunogen Recombinant protein within mouse EMC10 aa 1-258.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284361, 69683
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.