Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf70 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734941 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4941 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4941 Supplier Proteintech Group Inc Supplier No. 255141AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to C19orf70.

Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintece of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane.

Specifications

Antigen C19orf70
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MICOS13
Gene Accession No. Q5XKP0
Gene Alias C19orf70, MIC13, P117, Protein P117, Protein QIL1, QIL1
Gene Symbols Micos13
Host Species Rabbit
Immunogen C19orf70 Fusion Protein Ag22003
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125988, 224904
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.