Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1orf112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554884 Supplier Invitrogen™ Supplier No. PA554884
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCMFALLLAD RSWLLEQHTL EAFTQFAEGT NHEEIVPQCL SSEETKNKVV SFLEKTGFVD ETEAAKVERV KQEKGIFWEP F Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 68%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf112
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1orf112
Gene Accession No. Q9NSG2
Gene Alias C1orf112; chromosome 1 open reading frame 112; uncharacterized protein C1orf112
Gene Symbols C1orf112
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf112. Recombinant protein control fragment (Product #RP-93974).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55732
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.