Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7211794 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5588 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5588 Supplier Proteintech Group Inc Supplier No. 271751AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Component C2 is a serum glycoprotein that functions aspart of the classical pathway of the complement system. ActivatedC1 cleaves C2 into C2a and C2b. The serine proteinase C2a thencombines with complement factor 4b to create the C3 or C5convertase. Deficiency of C2 has been reported to associated withcertain autoimmune diseases and SNPs in this gene have beenassociated with altered susceptibility to age-related maculardegeneration. This gene localizes within the class III region ofthe MHC on the short arm of chromosome 6. Alternative splicingresults in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described in publicationsbut their full-length sequence has not been determined.

Specifications

Antigen C2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene C2
Gene Accession No. P06681
Gene Alias C2, C3/C5 convertase, CO2, Complement C2, complement component 2
Gene Symbols C2
Host Species Rabbit
Immunogen C2 Fusion Protein Ag25690
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 717
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.