Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf46 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7211053 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4486 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4486 Supplier Proteintech Group Inc Supplier No. 246781AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C20orf46, also named as TMEM74B, belongs to the TMEM74 family.

Specifications

Antigen C20orf46
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TMEM74B
Gene Accession No. Q9NUR3
Gene Alias C20orf46, Transmembrane protein C20orf46
Gene Symbols TMEM74B
Host Species Rabbit
Immunogen C20orf46 Fusion Protein Ag20385
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108832, 55321
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.