Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf34 Monoclonal Antibody (OTI5D1), TrueMAB™, OriGene
SDP

Catalog No. 501673487
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-3487 100 μL
50-254-1555 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-3487 Supplier OriGene Supplier No. TA502428
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MIR99AHG (Mir-99a-Let-7c Cluster Host Gene) is an RNA Gene, and is affiliated with the miRNA class. Diseases associated with MIR99AHG include Myeloid Leukemia.

Specifications

Antigen C21orf34
Applications Western Blot
Classification Monoclonal
Clone OTI5D1
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MIR99AHG
Gene Alias C21orf34, C21orf35, LINC00478, MONC
Gene Symbols MIR99AHG
Host Species Mouse
Immunogen Full length human recombit protein of human C21orf34 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388815
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.