Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf34 Monoclonal Antibody (OTI5D1), TrueMAB™, OriGene
SDP

Catalog No. p-200011026
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1555 30 μL
50-167-3487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1555 Supplier OriGene Supplier No. TA502428S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C21orf34 Monoclonal antibody specifically detects C21orf34 in Human samples. It is validated for Western Blot

Specifications

Antigen C21orf34
Applications Western Blot
Classification Monoclonal
Clone OTI5D1
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene UBL4A
Gene Accession No. P11441
Gene Alias C21orf34; C21orf35; LINC00478; MIR99AHG; mir-99a-let-7c cluster host gene; MONC
Host Species Mouse
Immunogen Full length human recombinant protein of human UBL4A produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8266
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.