Learn More
Invitrogen™ C21orf63 Polyclonal Antibody

Description
PA1-9538 detects C21 in human samples. PA1-9538 has been successfully used in Western blot procedures. By Western blot, PA1-9538 detects a 50 kDa protein representing C21. PA1-9538 immunogen is a purified recombinant C21 corresponding to the chromosome 21 gene (Human).
Specifications
Specifications
| Antigen | C21orf63 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | EVA1C |
| Gene Accession No. | P58658 |
| Gene Alias | 1700092M14Rik; 4931408A02Rik; B18; B19; C21; C21orf63; C21orf64; DC42; EVA1; EVA-1; eva-1 homolog C; eva-1 homolog C (C. elegans); EVA1C; Fam176c; family with sequence similarity 176, member C; FLJ12894; IRA1; PRED34; Protein eva-1 homolog C; protein FAM176C; RGD1307569; SUE21; TBLR1; UNQ2504/PRO5993 |
| Gene Symbols | EVA1C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.