Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C22orf18 Recombinant Protein

Catalog No. 89013474 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-013-474 10 μg
89-013-475 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-013-474 Supplier Abnova™ Supplier No. H00079019P0110
Add to Cart
Add to Cart

Human C22orf18 full-length ORF recombinant protein with GST-tag at N-terminal

The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3. CENPM is an additional factor required for centromere assembly.

  • Theoretical MW (kDa): 45.54
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MSVLRPLDKLPGLNTATILLVGTEDALLQQLADSMLKEDCASELKVHLAKSLPLPSSVNRPRIDLIVFVVNLHSKYSLQNTEESLRHVDASFFLGKVCFLATGAGRESHCSIHRHTVVKLAHTYQSPLLYCDLEVEGFRATMAQRLVRVLQICAGHVPGVSALNLLSLLRSSEGPSLEDL

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH00705
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 79019
Molecular Weight (g/mol) 45.54
Name C22orf18 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MSVLRPLDKLPGLNTATILLVGTEDALLQQLADSMLKEDCASELKVHLAKSLPLPSSVNRPRIDLIVFVVNLHSKYSLQNTEESLRHVDASFFLGKVCFLATGAGRESHCSIHRHTVVKLAHTYQSPLLYCDLEVEGFRATMAQRLVRVLQICAGHVPGVSALNLLSLLRSSEGPSLEDL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias C22orf18/CENP-M/MGC861/PANE1/bK250D10.2
Common Name CENPM
Gene Symbol CENPM
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.