Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ C26-A6
GREENER_CHOICE

Catalog No. p-7250135
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-921-0 10 mg
76-925-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-921-0 Supplier Tocris Bioscience™ Supplier No. 7692/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Disrupts MTDH and SND1 interaction, enhances immune surveillance

C26-A2 inhibits the interaction between MTDH and SND1 by blocking the SND1 pocket. C26-A2 disrupts MTDH-SND1 complex-induced reduction in tumor antigen presentation and T cell infiltration; enhancing immune surveillance and sensitivity to anti-PD-1 therapy in metastatic breast cancer mouse models. C26-A6 suppresses tumor growth (IC50 = 12.3 μM) and metastasis and enhances chemotherapy sensitivity in triple-negative breast cancer (TNBC) mouse models.

Specifications

Chemical Name or Material 5-Chloro-2-methoxy-N-(2-methyl[1,2,4 ]triazolo[1,5-a]pyridin-8-yl)benzenesulfonamide
CAS 1341030-00-9
Quantity 10 mg
Target Cell Adhesion Inhibitors
Molecular Formula C14H13ClN4O3S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.