Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf62 Monoclonal Antibody (OTI3G6), TrueMAB™, OriGene
SDP

Catalog No. p-200012771
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9654 30 μL
50-167-5975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9654 Supplier OriGene Supplier No. TA505509S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C2orf62 Monoclonal antibody specifically detects C2orf62 in Human samples. It is validated for Western Blot

Specifications

Antigen C2orf62
Applications Western Blot
Classification Monoclonal
Clone OTI3G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CATIP
Gene Accession No. Q7Z7H3
Gene Alias C15H2orf62; C2orf62; C37H2orf62; C6H2orf62; Catip; ciliogenesis associated TTC17 interacting protein; Ciliogenesis-associated TTC17-interacting protein; Gm216; hypothetical protein LOC690437; LOC690437
Host Species Mouse
Immunogen Full length human recombinant protein of human C2orf62 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375307
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.