Learn More
Aviva Systems Biology Corp C3 ELISA Kit (Mouse), 96 wells
SDP
Supplier:  Aviva Systems Biology Corp OKIA00089
Encompass_Preferred
Aviva Systems Biology C3 ELISA Kit (Mouse) is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody for Complement C3 has been pre-coated onto a 96-wellplate (12 x 8 Well Strips). Standards or test samples are added to the wells, incubated and washed to remove unbound proteins.
Catalog No. NC1785849
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.