Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C3 (Human) ELISA Kit

Catalog No. 89937500 Shop All Abnova Corporation Products
Change view
Click to view available options
Target Species:
Human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Species
89-937-500 Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-937-500 Supplier Abnova™ Supplier No. KA4797

Sandwich enzyme immunoassay for the quantitative measurement of human C3

  • Used for the in vitro quantitative determination of human complement C3 in human serum, human plasma, cell lysate and buffered solution
  • Assay will recognize native complement C3

Specifications

Accession Number P01024
Calibration Range 15.6 to 1000 ng/mL
Description C3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human C3.
Conjugate Biotin
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Buffered Solution, Cell Lysates, Plasma, Serum
Target Species Human
For Use With (Application) Quantification
Gene ID (Entrez) 718
Kit Contents Human C3 microtiter plate, Incubation Buffer, Washing Buffer (20x), Standard Protein, Standard/Sample Dilution Buffer, Secondary Antibody, AV-HRP, Secondary Antibody/AV-HRP Dilution Buffer, Substrate (Stabilized chromogen), Stop Solution: 1 N solution of sulfuric acid (H2SO4), Plate sealers: Adhesive sheet
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at 4°C.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.