Learn More
MyBioSource Porcine C3a ELISA Kit
SDP
Supplier:  MyBioSource MBS250936024 TESTS
Encompass_Preferred
The MyBioSource Porcine C3a ELISA Kit quantifies C3a in various biological fluids using a monoclonal antibody. It involves binding, washing, detection with a biotinylated antibody, and a streptavidin-HRP conjugate, followed by colorimetric measurement at 450 nm. The detection limit is <0.1 ng/mL.
Catalog No. NC2330255
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.