Learn More
Description
- C3b (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human C3b.
Specifications
Specifications
| Description | complement component 3 |
| Cross Reactivity | Human |
| Product Type | Kit |
| Gene ID (Entrez) | 718 |
| Gene Symbol | C3 |
| Quantity | 1 Kit |
| Storage Requirements | Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing. |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.