Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C3orf15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103598
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103598 100 μL
1 options

Catalog No. PIPA5103598

Supplier: Invitrogen™ PA5103598

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AAT1.

C3orf15 (MAATS1, MYCBP Associated And Testis Expressed 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include adenosylmethionine decarboxylase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAATS1
Gene Accession No. Q7Z4T9
Gene Alias 4932425I24Rik; AAT1; AAT-1; AAT1alpha; AAT1-alpha; AMY-1-associating protein expressed in testis 1; AMY-1-associating protein expressed in testis 1 homolog; C3orf15; CaM-IP2; CFAP91; cilia and flagella associated protein 91; cilia- and flagella-associated protein 91; Maats1; MYCBP associated and testis expressed 1; MYCBP/AMY-1-associated testis-expressed protein 1; MYCBP/AMY-1-associated, testis expressed 1; MYCBP-associated, testis expressed 1; MYCBP-binding protein; protein MAATS1; RGD1305539; SPATA26; spermatogenesis associated 26
Gene Symbols MAATS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAATS1(Accession Q7Z4T9), corresponding to amino acid residues C212-Q262.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89876
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.