Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf31 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735819 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5819 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5819 Supplier Proteintech Group Inc Supplier No. 281371AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C3orf31 may be involved in the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.

Specifications

Antigen C3orf31
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TAMM41
Gene Accession No. Q96BW9
Gene Alias C3orf31, DKFZp434E0519
Gene Symbols TAMM41
Host Species Rabbit
Immunogen C3orf31 Fusion Protein Ag26826
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132001
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.