Learn More
Alkali Scientific H3565-180HD 180L Plus CO2 Incu
SDP
Supplier:  Alkali Scientific RS7517
Encompass_Preferred
These new Alkali ScientificH3565-180HD 180L Plus CO2 Incubator for mammalian cell culture is loaded with features and options making them among the most versatile and safest cell culture incubators on the market The stainless steel interior delivers excellent temperature uniformity and has seamless rounded corners for quick and easy clean up The digital control panel precisely maintains the six-heating elements that surround the interior for unparalleled uniformity (/- 0 4C) With six-sided heating air-flow is minimized resulting in reduced concern for contamination Dual Infrared Beam sensors track and monitor the CO2 density accurately Transparent sealed inner glass doors allows for visualization of samples without exposing the chamber to ambient temperatures This improves performance and reduces energy waste The inner part of the door including the door frame is heated which helps reduce condensation from forming on the inner glass door The Alkali Scientific(H3565-180HD) 180L
Catalog No. 50-302-9532
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.