Learn More
Description
Mouse anti Human CD88 antibody, clone P12/1 recognizes the C5a receptor (C5aR) also known as CD88 or C5a anaphylatoxin chemotactic receptor 1. Reconstitute with 1.0 mL distilled water.
Specifications
Specifications
| Antigen | C5AR1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | P12/1 |
| Concentration | 1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 5% sucrose, 1% BSA and 0.09% sodium azide |
| Gene | C5AR1 |
| Gene Accession No. | P21730 |
| Gene Alias | anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
