Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5ORF44 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983555
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3555 Supplier Bioss Supplier No. BS15205R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of C5orf44 has not been widely studied, and is yet to be fully elucidated.

Specifications

Antigen C5ORF44
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Trappc13
Gene Accession No. A5PLN9
Gene Alias 2410002O22Rik; 2610524F24Rik; AI451892; AU017229; C5orf44; RGD1306583; trafficking protein particle complex 13; Trafficking protein particle complex subunit 13; TRAPPC13; Trs65-related; UPF0533 protein C5orf44; UPF0533 protein C5orf44 homolog
Gene Symbols Trappc13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C5ORF44.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80006
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.