Learn More
Estes™ Industries C6-5 Engines
SDPSCI_ED
Catalog No. S13199 Shop All Estes Industries Products
Encompass
Change view
Click to view available options
Includes: Everything needed to launch 24 rockets
Catalog No.
S13199 Includes: Everything needed to launch 24 rockets
1 options
This item is not returnable. View return policy
Catalog No. S13199 Supplier Estes™ Industries Supplier No. 1789
This item is not returnable. View return policy
C6-5 engine is the highest impulse engine in a bulk pack. This engine will send students' rockets out of site. Estes™ engine bulk packs are convenient and include everything your students need to prepare their rockets for launching 24 rockets.
  • Estes model rocket engine bulk packs are available in four total impulse ranges: ½A (mini engine), A, B or C (standard engines). These packs may be used for the rocket bulk packs or other Estes rockets. Whether you are launching rockets at school with students, at a youth group activity or rocket club monthly launch, Estes engine bulk packs are a must.
  • Engine bulk packs come with all the rocket engine supplies you need to launch 24 rockets. Included are 24 rocket engines, 30 model rocket starters, 24 reusable starter plugs and 75 squares of recovery wadding. Use the C6-5 engine to launch these bulk pack rockets: Alpha III, Generic E2X, Alpha, Viking and Wizard.
Specifications
Includes 24 x rocket engines, 30 x model rocket starters, 24 x reusable starter plugs, 75 x squares of recovery wadding
Product Type Model, Engine