Learn More
Avanti Polar Lipids C6 CERAMIDE POWDER 10 MG
Supplier:  Avanti Polar Lipids 860506P10MG
Encompass
N-Hexanoyl-D-erythro-Sphingosine (C6 Ceramide), (2S, 3R, 4E)-2-hexanoylaminooctadec-4-ene-1, 3-diol, (2S, 3R, 4E)-2-hexanoylamino-1, 3-octadec-4-enediol, C 24 H 47 NO3 Percent composition: C 72.49% H 11.91% N 3.52% O 12.07%, 10 Mg
Catalog No. NC9195706
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.