Learn More
BD Cell Analysis 3P C6 FLOW CYTOMETER FLUID KIT
Supplier:  BD Cell Analysis 3P 653158
Encompass
BD accuri Contents contains concentrate for making 1 Liter each of Cleaning and Decontamination Solutions and Bacteriostatic Concentrate for treating 10 L of Sheath; as well as 8mL of our Extended Flow Cell Clean. Solution application: Flow cytometry (Routinely Tested)
Catalog No. NC0356347