Learn More
Cayman Chemical Methyltrnsferase asy S-Ade 1ea
SDP
Supplier:  Cayman Chemical 7001461 EA
Encompass_Preferred
An anticancer polyamine inhibits the proliferation of PC3 TSU-pr1 DU145 and JCA-1 cells (IC50s 65 62 3 and 21 UM respectively) reduces ODC activity and increases diamine acetyltransferase 1 activity in PC3 DU145 and JCA-1 cells at 100 UM increases H2O2 levels and disrupts the mitochondrial membrane potential in U87 and LN-229 cells at 10 UM decreases tumor growth in a DU145 mouse xenograft model at 40 mg/kg three times per day
Catalog No. 50-289-4416
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.