Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf43 Monoclonal Antibody (OTI4F9), TrueMAB™, OriGene
SDP

Catalog No. p-200015388
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3678 Supplier OriGene Supplier No. TA810854
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C9orf43 is a protein coding gene.

Specifications

Antigen C9orf43
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene C9orf43
Gene Accession No. Q8TAL5
Gene Alias chromosome 9 open reading frame 43
Gene Symbols C9orf43
Host Species Mouse
Immunogen Full length human recombit protein of human C9ORF43 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257169
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.