Learn More
C9orf9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
Product No. 26034-1-AP is a rabbit polyclonal antibody raised against the full-length of human C9orf9. Two isoforms were produced by alternative splicing with predicted MW of 19 and 26 kDa. Two bands at 16 kDa and 30 kDa could be detected using the presen
Specifications
Specifications
| Antigen | C9orf9 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | SPACA9 |
| Gene Accession No. | Q7TPM5, Q96E40 |
| Gene Alias | C9orf9, Uncharacterized protein C9orf9 |
| Gene Symbols | Spaca9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.