Learn More
Siemens Healthineers CA Clean™ II Detergent
Catalog No. 23044640 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
45 mL
500 mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-640 5 L
23-044-744 45 mL
23-044-638 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-044-640 Supplier Siemens Healthineers Supplier No. 10874005
Add to Cart
Add to Cart
For use with Sysmex™ CA and CS Systems
CA Clean™ II detergent is used to cleanse the automated pipets used in Sysmex™ CA and CS systems.
TRUSTED_SUSTAINABILITY
Specifications
Type Detergent
For Use With (Application) Cleansing Automated Pipets
For Use With (Equipment) Sysmex™ CA & CS Systems
Quantity 5 L