Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ CA MG Hardness Solutions

Catalog No. 01670225 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
5 mL
15 mL
Type:
Ca/Mg Hardness Solution 1
Ca/Mg Hardness Solution 2
Ca/Mg Hardness Solution 3
Ca/Mg Hardness Solution 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
01-670-225 5 mL Ca/Mg Hardness Solution 3
01-670-223 15 mL Ca/Mg Hardness Solution 1
01-670-224 15 mL Ca/Mg Hardness Solution 2
01-670-226 5 mL Ca/Mg Hardness Solution 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 01-670-225 Supplier Lovibond™ Supplier No. 471230
Add to Cart
Add to Cart
This item is not returnable. View return policy

Liquid reagent used as part of a reagent set to determine the concentration of calcium and magnesium hardness in a water sample.

  • Ideal for analyzing water intended for pre-treatment of Reverse Osmosis (RO) and water softening systems, this reagent set is used to determine the concentration of calcium and magnesium hardness in a water sample.
  • Easy dispensing with dropper tip bottle
  • Liquid reagent dissolves and mixes easily into sample
  • Part of a simplified method for analyzing Calcium and Magnesium Hardness

Specifications

Analytes Calcium and Magnesium Hardness
Description Solution 3 of 4 in Lovibond's CA MG Hardness Set for measuring Calcium and Magnesium Hardness in a water sample.
For Use With (Application) Pre-Treatment of RO Water and Softening Systems
For Use With (Equipment) Lovibond Spectrophotometers and Colorimeters
Quantity 5 mL
Range 0.05 to 4 mg/L CaCO3
Test Method Calmagite
Type Ca/Mg Hardness Solution 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.