Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CA125 Monoclonal Antibody (X325)
GREENER_CHOICE

Catalog No. MA124603 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA124603 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA124603 Supplier Invitrogen™ Supplier No. MA124603
  • $639.00
    $518.65 / Each of 1

    Save $120.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-24603 detects MUC16 from human samples.

CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CA125
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone X325
Concentration 5.3 mg/mL
Conjugate Unconjugated
Formulation 50mM sodium citrate with 150mM NaCl and 0.09% sodium azide; pH 7.5
Gene MUC16
Gene Accession No. Q8WXI7
Gene Alias 1110008I14Rik; Ca125; CA-125; CA125 ovarian cancer antigen; ca-ovarian; FLJ14303; Gm1480; MUC16; MUC-16; mucin 16; mucin 16, cell surface associated; mucin-16; Ovarian cancer-related tumor marker CA125; ovarian carcinoma antigen CA125; RGD1308422
Gene Symbols MUC16
Host Species Mouse
Immunogen CA125 antigen purified from human ovarian carcinoma.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94025
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.